Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mctp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760610.1 | XP_008758832.1 | 8 | 1254 | ||
XM_008760611.1 | XP_008758833.1 | 7 | 1215 | |||
XM_008760612.1 | XP_008758834.1 | 9 | 3934 | |||
XM_008760613.1 | XP_008758835.1 | 8 | 3798 | |||
XM_008760614.1 | XP_008758836.1 | 8 | 1250 | |||
XM_008760615.1 | XP_008758837.1 | 7 | 1211 | |||
XM_008760616.1 | XP_008758838.1 | 4 | 3404 | |||
XM_008760617.1 | XP_008758839.1 | 4 | 285 | |||
GenBank mRNA | FQ232558.1 | - | 7 | 834 |