Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Car1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107660.1 | NP_001101130.1 | 4 | 339 | ||
XM_006232111.1 | XP_006232173.1 | 3 | 293 | |||
XM_008760842.1 | XP_008759064.1 | 4 | 296 | |||
GenBank mRNA | BC158888.1 | - | 4 | 302 | 8373855 | |
FQ225390.1 | - | 2 | 210 | |||
FQ225569.1 | - | 4 | 343 | |||
FQ225719.1 | - | 4 | 333 | |||
FQ231425.1 | - | 4 | 333 | |||
FQ233411.1 | - | 4 | 335 | |||
FQ235246.1 | - | 4 | 343 |