Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Egfem1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107663.1 | NP_001101133.1 | 10, 11 | 1456 | ||
XM_008760923.1 | XP_008759145.1 | 10, 11 | 1544 |