Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tspan18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107750.1 | NP_001101220.1 | 4 | 523 | ||
XM_006234573.2 | XP_006234635.1 | 4 | 454 | |||
XM_006234574.2 | XP_006234636.1 | 5 | 596 | |||
XM_006234575.2 | XP_006234637.1 | 3 | 374 | |||
XM_008762027.1 | XP_008760249.1 | 3 | 274 |