Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mtfr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100977.1 | NP_001094447.1 | 4 | 517 | ||
XM_006232178.2 | XP_006232240.1 | 5 | 674 | |||
XM_006232179.2 | XP_006232241.1 | 5 | 653 | |||
XM_006232180.2 | XP_006232242.1 | 4 | 1463 | |||
GenBank mRNA | EF221637.1 | - | 4 | 517 | ||
FQ217237.1 | - | 5 | 625 |