Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc52a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037198.1 | NP_001032275.1 | 4 | 1233 | ||
XM_006235238.2 | XP_006235300.1 | 7 | 1904 | |||
XM_008762327.1 | XP_008760549.1 | 7 | 2284 | |||
XM_008762328.1 | XP_008760550.1 | 4 | 2095 | |||
XM_008762329.1 | XP_008760551.1 | 3 | 1608 | |||
GenBank mRNA | BC099097.1 | - | 4 | 1233 | 7459579 |
Set Membership: |
G-protein coupled receptor |