Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107793.1 | NP_001101263.1 | 11 | 1425 | ||
XM_006235321.2 | XP_006235383.1 | 12 | 1532 | |||
GenBank mRNA | BC161963.1 | - | 11 | 1405 | 9028172 | |
FQ217492.1 | - | 12 | 1376 |