Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pmepa1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107807.2 | NP_001101277.1 | 3 | 488 | ||
XM_006235689.1 | XP_006235751.1 | 4 | 653 |