Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zbtb43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012094.1 | NP_001012094.1 | 3, 4 | 259 | ||
XM_008761749.1 | XP_008759971.1 | 1, 2 | 1286 | |||
GenBank mRNA | BC087042.1 | - | 3, 4 | 259 | 7107163 |