Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pex5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170584.1 | NP_001164055.1 | 4 | 414 | ||
XM_006237384.2 | XP_006237446.1 | 4 | 415 | |||
XM_006237385.2 | XP_006237447.1 | 4 | 345 | |||
GenBank mRNA | CK477113.1 | - | 4 | 283 | ||
FM036217.1 | - | 4 | 414 |
Gene Symbol: LOC103690024 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763277.1 | XP_008761499.1 | 346 |