Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014061.1 | NP_001014083.1 | 363 | |||
GenBank mRNA | BC079135.1 | - | 363 | 7127603 |
Gene Symbol: NEWGENE_1359502 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006237685.2 | XP_006237747.1 | 4 | 1776 | ||
XM_006237686.2 | XP_006237748.1 | 3 | 768 | |||
XM_006237687.2 | XP_006237749.1 | 3 | 768 | |||
XM_008763455.1 | XP_008761677.1 | 4 | 1776 | |||
XM_008763456.1 | XP_008761678.1 | 4 | 3150 |