Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col8a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006238912.2 | XP_006238974.1 | 4 | 1786 | ||
XM_233542.7 | XP_233542.4 | 4 | 1780 |