Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrpprc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008519.1 | NP_001008519.1 | 31 | 3351 | ||
GenBank mRNA | AY293808.1 | - | 31 | 3351 |