Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rasgrp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108009.1 | NP_001101479.1 | 6 | 580 | ||
XM_006239757.2 | XP_006239819.1 | 6 | 580 | |||
XM_006239758.2 | XP_006239820.1 | 7 | 1017 | |||
XM_006239759.2 | XP_006239821.1 | 6 | 909 | |||
XM_006239760.2 | XP_006239822.1 | 6 | 884 | |||
XM_006239761.2 | XP_006239823.1 | 4 | 405 | |||
XM_006239762.2 | XP_006239824.1 | 5 | 776 | |||
XM_008764485.1 | XP_008762707.1 | 2 | 1075 |