Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mdm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108099.1 | NP_001101569.1 | 11 | 1351 | ||
XM_006241382.1 | XP_006241444.1 | 12 | 1716 | |||
XM_006241383.1 | XP_006241445.1 | 12 | 1280 | |||
XM_008765396.1 | XP_008763618.1 | 13 | 1788 | |||
XM_008765397.1 | XP_008763619.1 | 13 | 1352 | |||
XM_008765399.1 | XP_008763621.1 | 9 | 1098 |