Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apobec3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033703.1 | NP_001028875.1 | 4 | 647 | ||
XM_008765715.1 | XP_008763937.1 | 4 | 1730 | |||
XM_008765716.1 | XP_008763938.1 | 4 | 1783 | |||
GenBank mRNA | BC101878.1 | - | 4 | 647 | 7929998 |