Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Jam3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004269.1 | NP_001004269.1 | 4, 5 | 416 | ||
XM_008766054.1 | XP_008764276.1 | 3, 4 | 905 | |||
GenBank mRNA | BC079429.1 | - | 4, 5 | 416 | 7114667 |