Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp49 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136470.1 | NP_001129942.1 | 6 | 2252 | ||
XM_006244435.2 | XP_006244497.1 | 5 | 1855 | |||
XM_008766857.1 | XP_008765079.1 | 4 | 1973 | |||
XM_008766858.1 | XP_008765080.1 | 5 | 2351 | |||
XM_008766859.1 | XP_008765081.1 | 4 | 1973 |