Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gtf2a1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012136.1 | NP_001012136.1 | 7 | 1165 | ||
XM_006239740.2 | XP_006239802.1 | 8 | 1855 | |||
XM_006239741.2 | XP_006239803.1 | 7 | 1754 | |||
XM_008764478.1 | XP_008762700.1 | 10 | 1718 | |||
GenBank mRNA | BC082104.1 | - | 7 | 1165 | 7134703 |