Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdr2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006247771.2 | XP_006247833.1 | 4 | 885 | ||
XM_006247772.1 | XP_006247834.1 | 4 | 784 | |||
XM_008768410.1 | XP_008766632.1 | 4 | 604 | |||
XM_340933.7 | XP_340934.5 | 4 | 886 |