Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014792.1 | NP_001014792.1 | 5, 6 | 284 | ||
XM_006252127.2 | XP_006252189.1 | 5, 6 | 378 | |||
XM_008770761.1 | XP_008768983.1 | 5, 6 | 233 | |||
GenBank mRNA | AY779027.1 | - | 5, 6 | 284 |