Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEWGENE_1305243 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_341690.7 | XP_341691.7 | 1581 | |||
GenBank mRNA | FQ230829.1 | - | 1533 |