Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nrm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212508.2 | NP_997673.1 | 2 | 236 | ||
XM_008772732.1 | XP_008770954.1 | 2 | 1168 | |||
GenBank mRNA | BC098637.1 | - | 2 | 197 | ||
BP504258.1 | - | 2 | 242 | |||
FQ220138.1 | - | 2 | 194 |