Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Atp8b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024798.2 | NP_001019969.2 | 14 | 1485 | ||
XM_006232697.2 | XP_006232759.1 | 14 | 1543 | |||
XM_006232699.2 | XP_006232761.1 | 14 | 1528 | |||
XM_006232702.2 | XP_006232764.1 | 13 | 1435 | |||
XM_006232706.1 | XP_006232768.1 | 12 | 1333 | |||
XM_008761181.1 | XP_008759403.1 | 14 | 1496 | |||
XM_008761182.1 | XP_008759404.1 | 14 | 1543 | |||
XM_008761183.1 | XP_008759405.1 | 13 | 1468 | |||
XM_008761184.1 | XP_008759406.1 | 12 | 1366 | |||
GenBank mRNA | BC097468.1 | - | 14 | 1518 |