Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ahcyl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108561.1 | NP_001102031.1 | 3 | 355 | ||
XM_006233165.1 | XP_006233227.1 | 2 | 239 | |||
XM_006233166.2 | XP_006233228.1 | 3 | 579 | |||
XM_006233167.1 | XP_006233229.1 | 2 | 239 | |||
XM_008761405.1 | XP_008759627.1 | 3 | 591 | |||
XM_008761406.1 | XP_008759628.1 | 3 | 591 | |||
GenBank mRNA | BC169126.1 | - | 2 | 219 |