Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stxbp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191872.1 | NP_001178801.1 | 4 | 715 | ||
XM_006240067.2 | XP_006240129.1 | 4 | 2277 | |||
XM_008764658.1 | XP_008762880.1 | 5 | 780 | |||
XM_008764659.1 | XP_008762881.1 | 4 | 732 |