Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lin52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006240356.2 | XP_006240418.1 | 4 | 657 | ||
XM_006240358.2 | XP_006240420.1 | 4 | 483 | |||
XM_008764889.1 | XP_008763111.1 | 4 | 480 | |||
XM_008764890.1 | XP_008763112.1 | 4 | 473 | |||
XM_343089.7 | XP_343090.4 | 4 | 469 |