Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arhgap9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012198.1 | NP_001012198.1 | 9 | 743 | ||
NM_001080789.1 | NP_001074258.1 | 10 | 1375 | |||
XM_006241464.2 | XP_006241526.1 | 10 | 1462 | |||
XM_006241465.2 | XP_006241527.1 | 10 | 1453 | |||
XM_006241466.2 | XP_006241528.1 | 10 | 1496 | |||
XM_006241467.2 | XP_006241529.1 | 10 | 1506 | |||
XM_006241468.2 | XP_006241530.1 | 9 | 800 | |||
GenBank mRNA | BC079334.1 | - | 9 | 743 | 7097179 | |
BC107938.1 | - | 10 | 1375 | 7378387 |