Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tcaim | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110838.1 | NP_001104308.1 | 5 | 504 | ||
XM_006244192.2 | XP_006244254.1 | 5 | 616 | |||
XM_006244193.2 | XP_006244255.1 | 5 | 725 | |||
XM_006244194.2 | XP_006244256.1 | 5 | 641 | |||
XM_006244195.2 | XP_006244257.1 | 5 | 609 | |||
XM_008766717.1 | XP_008764939.1 | 5 | 641 | |||
XM_008766718.1 | XP_008764940.1 | 4 | 750 | |||
XM_008766719.1 | XP_008764941.1 | 4 | 508 | |||
XM_008766720.1 | XP_008764942.1 | 4 | 473 | |||
XM_008766721.1 | XP_008764943.1 | 4 | 505 | |||
XM_008766722.1 | XP_008764944.1 | 5 | 615 | |||
XM_008766723.1 | XP_008764945.1 | 4 | 620 | |||
GenBank mRNA | FQ210480.1 | - | 5 | 518 | ||
FQ211645.1 | - | 5 | 589 | |||
FQ215070.1 | - | 5 | 518 |