Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prss39 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013226.1 | NP_001013244.1 | 3 | 693 | ||
XM_006244717.2 | XP_006244779.1 | 4 | 749 | |||
XM_006244718.2 | XP_006244780.1 | 4 | 377 | |||
GenBank mRNA | BC079254.1 | - | 3 | 693 | 7133201 |