Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cops8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013227.2 | NP_001013245.1 | 1 | 110 | ||
GenBank mRNA | BC063182.1 | - | 1 | 114 | 6887869 | |
FQ215552.1 | - | 1 | 109 |