Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1362 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006598.1 | NP_001006598.1 | 1 | 422 | ||
GenBank mRNA | AF091566.1 | - | 1 | 233 |