Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108868.1 | NP_001102338.2 | 20 | 3167 | ||
XM_006252000.2 | XP_006252062.1 | 20 | 3865 | |||
GenBank mRNA | BC169074.1 | - | 12 | 1405 |