Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ticam2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108890.1 | NP_001102360.1 | 3 | 876 | ||
XM_006254690.2 | XP_006254752.1 | 2 | 949 |