Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1309821 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108971.1 | NP_001102441.1 | 3 | 2304 | ||
XM_006238094.2 | XP_006238156.1 | 2 | 2323 | |||
XM_008763637.1 | XP_008761859.1 | 2 | 2508 |