Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108976.1 | NP_001102446.1 | 2 | 348 | ||
XM_006238895.2 | XP_006238957.1 | 2 | 373 | |||
XM_006238896.2 | XP_006238958.1 | 2 | 1181 | |||
XM_006238897.2 | XP_006238959.1 | 2 | 366 | |||
GenBank mRNA | BC166902.1 | - | 2 | 308 | 6922293 |