Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zdhhc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039340.1 | NP_001034429.1 | 10 | 1065 | ||
GenBank mRNA | AY886533.1 | - | 10 | 1065 |