Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rac2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008384.1 | NP_001008385.1 | 6 | 622 | ||
XM_006242028.2 | XP_006242090.1 | 6 | 665 | |||
GenBank mRNA | BC086399.1 | - | 6 | 622 | 7303112 | |
FQ213630.1 | - | 6 | 621 | |||
FQ225192.1 | - | 6 | 626 | |||
FQ230979.1 | - | 6 | 664 | |||
FQ232209.1 | - | 6 | 646 | |||
FQ234815.1 | - | 6 | 646 | |||
FQ234871.1 | - | 6 | 599 |