Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col6a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008767340.1 | XP_008765562.1 | 4 | 1364 | ||
XM_008767341.1 | XP_008765563.1 | 4 | 1364 | |||
XM_008767342.1 | XP_008765564.1 | 4 | 1363 | |||
XM_008767343.1 | XP_008765565.1 | 3 | 742 |