Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptges3l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014272.1 | NP_001014294.1 | 2 | 663 | ||
GenBank mRNA | BC087125.1 | - | 2 | 663 | 7110309 |
Gene Symbol: Ptges3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130989.1 | NP_001124461.1 | 5 | 594 | ||
XM_008765026.1 | XP_008763248.1 | 5 | 637 | |||
GenBank mRNA | BC166579.1 | - | 5 | 594 | 7379926 |