Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199119.1 | NP_954550.1 | 5 | 1511 | ||
XM_006240483.1 | XP_006240545.1 | 5 | 1714 | |||
XM_006240484.2 | XP_006240546.1 | 5 | 1946 | |||
GenBank mRNA | AY341876.1 | - | 5 | 1511 | ||
BC097262.1 | - | 5 | 1498 | 7442976 | ||
FQ212095.1 | - | 3 | 257 |
Gene Symbol: LOC100909476 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750226.3 | XP_003750274.3 | 2 | 840 |