Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Taf9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037310.1 | NP_001032387.1 | 3 | 909 | ||
NM_184048.2 | NP_908937.2 | 3 | 430 | |||
GenBank mRNA | AY325235.1 | - | 2 | 266 | ||
BC091109.1 | - | 3 | 419 | 7322630 | ||
CB724309.1 | - | 3 | 419 | |||
FQ213120.1 | - | 3 | 413 | |||
FQ215462.1 | - | 3 | 424 | |||
FQ229736.1 | - | 3 | 423 |