Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1291 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000797.1 | NP_001000797.1 | 1 | 943 |
Gene Symbol: LOC100911064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751484.1 | XP_003751532.1 | 1 | 943 |