Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1253 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000805.1 | NP_001000805.1 | 1 | 582 |
Gene Symbol: LOC100912543 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762453.1 | XP_008760675.1 | 1 | 582 |
Gene Symbol: LOC103692041 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008762675.1 | XP_008760897.1 | 1 | 582 |