Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RT1-T18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001002821.1 | NP_001002821.1 | 358 | |||
XM_006255985.2 | XP_006256047.1 | 2, 3 | 925 | |||
XM_006255986.2 | XP_006256048.1 | 2, 3 | 925 | |||
XM_008772742.1 | XP_008770964.1 | 2, 3 | 925 | |||
XM_008772743.1 | XP_008770965.1 | 3, 4 | 390 | |||
XM_008772744.1 | XP_008770966.1 | 3, 4 | 1528 | |||
GenBank mRNA | AY144125.1 | - | 279 | |||
BC104719.1 | - | 763 |
Gene Symbol: LOC102552616 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256027.2 | XP_006256089.1 | 2, 3 | 654 |