Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008814.1 | NP_001008814.1 | 9 | 1604 | ||
XM_008765774.1 | XP_008763996.1 | 9 | 1653 | |||
GenBank mRNA | AB355638.1 | - | 9 | 1607 |