Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rnf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109025.2 | NP_001102495.1 | 3 | 323 | ||
GenBank mRNA | BC088341.1 | - | 3 | 299 | 7367801 | |
DY573537.1 | - | 3 | 323 |