Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcst | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005900.1 | NP_001005900.1 | 3 | 206 | ||
XM_006228785.2 | XP_006228847.1 | 3 | 470 | |||
GenBank mRNA | AY247020.1 | - | 3 | 206 |