Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rcn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008694.2 | NP_001008694.1 | 4 | 581 | ||
GenBank mRNA | BC083719.1 | - | 4 | 582 | 7190838 | |
CB315609.1 | - | 1 | 31 | |||
CK601329.1 | - | 4 | 581 | |||
FQ228883.1 | - | 4 | 601 |