Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klra2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009718.1 | NP_001009718.1 | 7 | 808 | ||
XM_008763403.1 | XP_008761625.1 | 6 | 870 | |||
GenBank mRNA | AY651018.1 | - | 7 | 808 |
Gene Symbol: Klra5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008763344.1 | XP_008761566.1 | 1011 |